Assay Details
| Gene Symbol: ITLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101275.1 | NP_001094745.1 | 2 - 3 | 131 | 76 | |
| GenBank mRNA | AM749043.1 | - | 2 - 3 | 131 | 76 | |
More Information
Set Membership: |
|

ITLN2
Genomic Map