Assay Details
| Gene Symbol: TOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102096.2 | NP_001095566.1 | 3 - 4 | 552 | 60 | |
| GenBank mRNA | BC147899.1 | - | 3 - 4 | 533 | 60 | |
| EH164370.1 | - | 3 - 4 | 552 | 60 | ||
More Information
Set Membership: |
|

TOX
Genomic Map