Assay Details
| Gene Symbol: MAP1S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102114.1 | NP_001095584.1 | 2 - 3 | 263 | 52 | |
| GenBank mRNA | BC149678.1 | - | 2 - 3 | 263 | 52 | |
More Information
Set Membership: |
|

MAP1S
Genomic Map