Assay Details
| Gene Symbol: DNAJA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102120.2 | NP_001095590.2 | 6 - 7 | 1132 | 69 | |
| GenBank mRNA | BC110008.1 | - | 6 - 7 | 795 | 69 | |
| BC151654.1 | - | 6 - 7 | 908 | 69 | ||
| DN511895.1 | - | 2 - 3 | 372 | 69 | ||
More Information
Set Membership: |
|

DNAJA4
Genomic Map