Assay Details
| Gene Symbol: FRMD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102133.1 | NP_001095603.1 | 15 - 16 | 2011 | 59 | |
| GenBank mRNA | BC148070.1 | - | 15 - 16 | 2011 | 59 | |
More Information
Set Membership: |
|

FRMD6
Genomic Map