Assay Details
| Gene Symbol: SP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102142.1 | NP_001095612.1 | 1 - 2 | 45 | 86 | |
| GenBank mRNA | BC151270.1 | - | 1 - 2 | 45 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151270.1, NM_001102142.1 transcripts.
Set Membership: |
|

SP7
Genomic Map