Assay Details
| Gene Symbol: PTCHD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102143.2 | NP_001095613.1 | 1 | 741 | 72 | |
| GenBank mRNA | BC149383.1 | - | 1 | 742 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTCHD4| Gene Symbol: PTCHD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102143.2 | NP_001095613.1 | 1 | 741 | 72 | |
| GenBank mRNA | BC149383.1 | - | 1 | 742 | 72 | |
Set Membership: |
|