Assay Details
| Gene Symbol: FGFR1OP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102147.1 | NP_001095617.1 | 3 - 4 | 431 | 83 | |
| GenBank mRNA | BC148973.1 | - | 3 - 4 | 431 | 83 | |
More Information
Set Membership: |
|

FGFR1OP2
Genomic Map