Assay Details
| Gene Symbol: RAB40B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102166.2 | NP_001095636.1 | 1 - 2 | 281 | 58 | |
| GenBank mRNA | BC150037.1 | - | 1 - 2 | 282 | 58 | |
More Information
Set Membership: |
|

RAB40B
Genomic Map