Assay Details
| Gene Symbol: CCN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102176.1 | NP_001095646.1 | 1 - 2 | 77 | 147 | |
| GenBank mRNA | BT030745.1 | - | 1 - 2 | 77 | 147 | |
More Information
Set Membership: |
|

CCN5
Genomic Map