Assay Details
| Gene Symbol: PBK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102178.1 | NP_001095648.1 | 4 - 5 | 467 | 159 | |
| GenBank mRNA | BC112889.1 | - | 4 - 5 | 490 | 159 | |
| BC150025.1 | - | 4 - 5 | 467 | 159 | ||
More Information
Set Membership: |
|

PBK
Genomic Map