Assay Details
| Gene Symbol: ERG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102183.1 | NP_001095653.1 | 7 - 8 | 840 | 70 | |
| GenBank mRNA | BC148054.1 | - | 7 - 8 | 840 | 70 | |
More Information
Set Membership: |
|

ERG
Genomic Map