Assay Details
| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102200.1 | NP_001095670.1 | 1 - 2 | 111 | 83 | |
| GenBank mRNA | BC150038.1 | - | 1 - 2 | 111 | 83 | |
More Information
Set Membership: |
|

THAP8
Genomic Map