Assay Details
| Gene Symbol: PANX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102225.1 | NP_001095695.1 | 1 - 2 | 794 | 66 | |
| GenBank mRNA | BC150089.1 | - | 1 - 2 | 794 | 66 | |
More Information
Set Membership: |
|

PANX2
Genomic Map