Assay Details
| Gene Symbol: TPGS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102244.2 | NP_001095714.1 | 2 - 3 | 202 | 65 | |
| GenBank mRNA | BC149011.1 | - | 2 - 3 | 204 | 65 | |
More Information
Set Membership: |
|

TPGS2
Genomic Map