Assay Details
| Gene Symbol: NOP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102257.1 | NP_001095727.1 | 11 - 12 | 1309 | 63 | |
| GenBank mRNA | BC149978.1 | - | 11 - 12 | 1309 | 63 | |
| BC151441.1 | - | 11 - 12 | 1309 | 63 | ||
More Information
Set Membership: |
|

NOP2
Genomic Map