Assay Details
| Gene Symbol: GBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102261.2 | NP_001095731.1 | 10 - 11 | 1755 | 124 | |
| GenBank mRNA | BC149507.1 | - | 10 - 11 | 1757 | 124 | |
More Information
Set Membership: |
|

GBP4
Genomic Map