Assay Details
| Gene Symbol: GBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102261.2 | NP_001095731.1 | 1 - 2 | 97 | 55 | |
| GenBank mRNA | BC149507.1 | - | 1 - 2 | 99 | 55 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149507.1, NM_001102261.2 transcripts.
Set Membership: |
|

GBP4
Genomic Map