Assay Details
| Gene Symbol: FMR1NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102262.2 | NP_001095732.1 | 3 - 4 | 647 | 61 | |
| GenBank mRNA | BC149653.1 | - | 3 - 4 | 648 | 61 | |
More Information
Set Membership: |
|

FMR1NB
Genomic Map