Assay Details
| Gene Symbol: UHRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102270.1 | NP_001095740.1 | 11 - 12 | 1882 | 63 | |
| GenBank mRNA | BC148949.1 | - | 11 - 12 | 1882 | 63 | |
More Information
Set Membership: |
|

UHRF2
Genomic Map