Assay Details
| Gene Symbol: BABAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102274.2 | NP_001095744.1 | 5 - 6 | 581 | 75 | |
| GenBank mRNA | BC150022.1 | - | 5 - 6 | 583 | 75 | |
More Information
Set Membership: |
|

BABAM2
Genomic Map