Assay Details
| Gene Symbol: TRAPPC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102290.1 | NP_001095760.1 | 2 - 3 | 1276 | 62 | |
| GenBank mRNA | BC147918.1 | - | 2 - 3 | 1276 | 62 | |
More Information
Set Membership: |
|

TRAPPC12
Genomic Map