Assay Details
| Gene Symbol: STPG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102304.2 | NP_001095774.1 | 6 - 7 | 709 | 58 | |
| GenBank mRNA | BC149661.1 | - | 6 - 7 | 710 | 58 | |
More Information
Set Membership: |
|

STPG1
Genomic Map