Assay Details
| Gene Symbol: COMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102317.1 | NP_001095787.1 | 2 - 3 | 352 | 67 | |
| GenBank mRNA | BC151585.1 | - | 2 - 3 | 352 | 67 | |
More Information
Set Membership: |
|

COMT
Genomic Map