Assay Details
| Gene Symbol: NARF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102318.1 | NP_001095788.1 | 5 - 6 | 590 | 70 | |
| GenBank mRNA | BC148983.1 | - | 5 - 6 | 590 | 70 | |
More Information
Set Membership: |
|

NARF
Genomic Map