Assay Details
| Gene Symbol: PLA2G2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102522.1 | NP_001095992.1 | 5 | 3114 | 113 | |
| GenBank mRNA | BC147912.1 | - | 5 | 3114 | 113 | |
More Information
Set Membership: |
|

PLA2G2F
Genomic Map