Assay Details
| Gene Symbol: SPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103102.2 | NP_001096572.1 | 1 - 2 | 65 | 110 | |
| GenBank mRNA | BC149936.1 | - | 1 - 2 | 66 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149936.1, NM_001103102.2 transcripts.
Set Membership: |
|

SPN
Genomic Map