Assay Details
| Gene Symbol: SUGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103278.1 | NP_001096748.1 | 10 - 11 | 1501 | 59 | |
| GenBank mRNA | BC123792.1 | - | 10 - 11 | 1501 | 59 | |
More Information
Set Membership: |
|

SUGP1
Genomic Map