Assay Details
| Gene Symbol: CIP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103284.2 | NP_001096754.1 | 18 - 19 | 2604 | 103 | |
| GenBank mRNA | BC134461.1 | - | 18 - 19 | 2605 | 103 | |
More Information
Set Membership: |
|

CIP2A
Genomic Map