Assay Details
| Gene Symbol: GTF2E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103294.1 | NP_001096764.1 | 1 - 2 | 30 | 82 | |
| GenBank mRNA | BC147980.1 | - | 1 - 2 | 30 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC147980.1, NM_001103294.1 transcripts.
Set Membership: |
|

GTF2E1
Genomic Map