Assay Details
| Gene Symbol: AP1S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103296.2 | NP_001096766.1 | 4 - 5 | 563 | 85 | |
| GenBank mRNA | BC142333.1 | - | 4 - 5 | 564 | 85 | |
More Information
Set Membership: |
|

AP1S3
Genomic Map