Assay Details
| Gene Symbol: PTP4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104998.1 | NP_001098468.1 | 3 - 4 | 599 | 67 | |
| GenBank mRNA | BC147865.1 | - | 3 - 4 | 599 | 67 | |
| BC151453.1 | - | 3 - 4 | 599 | 67 | ||
More Information
Set Membership: |
|

PTP4A2
Genomic Map