Assay Details
| Gene Symbol: CYB561D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105398.1 | NP_001098868.1 | 2 - 3 | 221 | 73 | |
| GenBank mRNA | BC134607.1 | - | 2 - 3 | 221 | 73 | |
More Information
Set Membership: |
|

CYB561D1
Genomic Map