Assay Details
| Gene Symbol: MAGEB16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105399.2 | NP_001098869.1 | 2 - 3 | 387 | 74 | |
| GenBank mRNA | BC149683.1 | - | 2 - 3 | 388 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149683.1, NM_001105399.2 transcripts.
Set Membership: |
|

MAGEB16
Genomic Map