Assay Details
| Gene Symbol: SH2D2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105401.2 | NP_001098871.1 | 3 - 4 | 439 | 69 | |
| GenBank mRNA | BC153239.1 | - | 3 - 4 | 440 | 69 | |
| BT026286.1 | - | 2 - 3 | 336 | 69 | ||
More Information
Set Membership: |
|

SH2D2A
Genomic Map