Assay Details
| Gene Symbol: RPP40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105402.2 | NP_001098872.1 | 7 - 8 | 910 | 62 | |
| GenBank mRNA | BC134788.1 | - | 7 - 8 | 912 | 62 | |
More Information
Set Membership: |
|

RPP40
Genomic Map