Assay Details
| Gene Symbol: WASHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105406.1 | NP_001098876.1 | 1 - 2 | 194 | 125 | |
| GenBank mRNA | BC153261.1 | - | 1 - 2 | 194 | 125 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC153261.1, NM_001105406.1 transcripts.
Set Membership: |
|

WASHC1
Genomic Map