Assay Details
| Gene Symbol: GPRC5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105412.1 | NP_001098882.1 | 1 - 2 | 48 | 84 | |
| GenBank mRNA | BC146089.1 | - | 1 - 2 | 48 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC146089.1, NM_001105412.1 transcripts.
Set Membership: |
|

GPRC5D
Genomic Map