Assay Details
| Gene Symbol: GPRC5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105412.1 | NP_001098882.1 | 2 - 3 | 989 | 73 | |
| GenBank mRNA | BC146089.1 | - | 2 - 3 | 989 | 73 | |
More Information
Set Membership: |
|

GPRC5D
Genomic Map