Assay Details
| Gene Symbol: FYB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105414.1 | NP_001098884.1 | 8 - 9 | 1861 | 58 | |
| GenBank mRNA | BC134622.1 | - | 8 - 9 | 1861 | 58 | |
More Information
Set Membership: |
|

FYB1
Genomic Map