Assay Details
| Gene Symbol: DDX28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105422.2 | NP_001098892.1 | 1 | 1645 | 73 | |
| GenBank mRNA | BC134724.1 | - | 1 | 1646 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DDX28| Gene Symbol: DDX28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105422.2 | NP_001098892.1 | 1 | 1645 | 73 | |
| GenBank mRNA | BC134724.1 | - | 1 | 1646 | 73 | |
Set Membership: |
|