Assay Details
| Gene Symbol: HHEX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105424.1 | NP_001098894.1 | 3 - 4 | 617 | 84 | |
| GenBank mRNA | BC134632.1 | - | 3 - 4 | 617 | 84 | |
| EU360978.1 | - | 2 - 3 | 178 | 84 | ||
More Information
Set Membership: |
|

HHEX
Genomic Map