Assay Details
| Gene Symbol: AUH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105436.2 | NP_001098906.1 | 8 - 9 | 876 | 64 | |
| GenBank mRNA | BC114832.1 | - | 8 - 9 | 877 | 64 | |
More Information
Set Membership: |
|

AUH
Genomic Map