Assay Details
| Gene Symbol: C24H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105439.1 | NP_001098909.1 | 3 - 4 | 1220 | 146 | |
| GenBank mRNA | BC142242.1 | - | 3 - 4 | 1220 | 146 | |
More Information
Set Membership: |
|

C24H18orf25
Genomic Map