Assay Details
| Gene Symbol: INKA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105446.1 | NP_001098916.1 | 1 - 2 | 183 | 114 | |
| GenBank mRNA | BC147959.1 | - | 1 - 2 | 183 | 114 | |
More Information
Set Membership: |
|

INKA1
Genomic Map