Assay Details
| Gene Symbol: AK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105608.2 | NP_001099078.1 | 4 - 5 | 349 | 164 | |
| GenBank mRNA | BC142021.1 | - | 4 - 5 | 350 | 164 | |
More Information
Set Membership: |
|

AK6
Genomic Map