Assay Details
| Gene Symbol: WDR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105623.1 | NP_001099093.1 | 1 - 2 | 107 | 100 | |
| GenBank mRNA | BC153250.1 | - | 1 - 2 | 107 | 100 | |
More Information
Set Membership: |
|

WDR6
Genomic Map