Assay Details
| Gene Symbol: VCPKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110083.2 | NP_001103553.1 | 3 - 4 | 468 | 141 | |
| GenBank mRNA | BC149565.1 | - | 3 - 4 | 469 | 141 | |
More Information
Set Membership: |
|

VCPKMT
Genomic Map