Assay Details
| Gene Symbol: HBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110507.1 | NP_001103977.1 | 2 - 3 | 391 | 80 | |
| GenBank mRNA | DN280438.1 | - | 2 - 3 | 338 | 80 | |
| DN526838.1 | - | 2 - 3 | 401 | 80 | ||
More Information
Set Membership: |
|

HBE1
Genomic Map