Assay Details
Gene Symbol: KCNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113281.1 | NP_001106752.1 | 3 - 4 | 1720 | 96 | |
GenBank mRNA | DN524278.1 | - | 1 - 2 | 463 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
